Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGCS2 Rabbit anti-Human, Mouse, Rat, Clone: 5T5T3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | HMGCS2 |
---|---|
Clone | 5T5T3 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
HMGCS2 Monoclonal antibody specifically detects HMGCS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HMGCS2 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3-hydroxy-3-methylglutaryl coenzyme A synthase, 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial), 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial), EC 2.3.3.10, HMG-CoA synthase, hydroxymethylglutaryl-CoA synthase, mitochondrial, mitochondrial 3-hydroxy-3-methylglutaryl-CoA synthase | |
A synthetic peptide corresponding to a sequence within amino acids 409-508 of human HMGCS2 (P54868). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5T5T3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3158 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title