Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGCS2 Recombinant Rabbit Monoclonal Antibody (5T5T3)
GREENER_CHOICE

Catalog No. PIMA537869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537869 Supplier Invitrogen™ Supplier No. MA537869
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, Mouse liver, Rat liver Immunogen sequence: AFSYGSGLAA SFFSFRVSQD AAPGSPLDKL VSSTSDLPKR LASRKCVSPE EFTEIMNQRE QFYHKVNFSP PGDTNSLFPG TWYLERVDEQ HRRKYARRPV.

The protein encoded by this gene belongs to the HMG-CoA synthase family. It is a mitochondrial enzyme that catalyzes the first reaction of ketogenesis, a metabolic pathway that provides lipid-derived energy for various organs during times of carbohydrate deprivation, such as fasting. Mutations in this gene are associated with HMG-CoA synthase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGCS2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 5T5T3
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HMGCS2
Gene Accession No. P22791, P54868, P54869
Gene Alias 1300002P16; 3-hydroxy-3-methylglutary-Coenzyme A synthase 2; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 2; 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial); 3-hydroxy-3-methylglutaryl-coenzyme A synthase; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial); hmgCoA; HMG-CoA synthase; Hmgcs1; Hmgcs2; hydroxymethylglutaryl-CoA synthase 2; hydroxymethylglutaryl-CoA synthase, mitochondrial; mHS; Mt3h3mg; testicular tissue protein Li 88
Gene Symbols HMGCS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 409-508 of human HMGCS2 (P54868).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15360, 24450, 3158
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.