| Antigen |
HMGN1/HMG14 (p Ser20, p Ser24) |
| Applications |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification |
Polyclonal |
| Concentration |
1.2 mg/mL |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 1:500-1:2000, ELISA 1:5000-1:25000, Immunohistochemistry 20 ug/ml, Immunohistochemistry-Paraffin 20 ug/ml |
| Formulation |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. |
P05114 |
| Gene Alias |
FLJ31471, High mobility group nucleosome-binding domain-containing protein 1, high-mobility group (nonhistone chromosomal) protein 14, high-mobility group nucleosome binding 1, high-mobility group nucleosome binding domain 1, HMG14FLJ27265, MGC104230, MGC117425, non-histone chromosomal protein HMG-14, nonhistone chromosomal protein HMG-14 |
| Gene Symbols |
HMGN1 |
| Host Species |
Rabbit |
| Immunogen |
This affinity purified HMGN1/HMG14 [p Ser20, p Ser24] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 19-28 of human HMGN1/HMG141/HMG14 protein (see below). (Uniprot: P05114) |
| Molecular Weight of Antigen |
10.66 kDa |
| Purification Method |
Affinity Purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
3150 |
| Test Specificity |
This affinity-purified antibody is directed against phosphorylated human HMGN protein at pS20 and pS24 residues (other HMGN proteins have this conserved sequence but at other residue positions). The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human HMGN pS20/pS24 proteins and the antibody is specific for the phosphorylated form of the proteins. Reactivity is seen against HMGN1 and HMGN2. HMGN3 is also likely reactive although not tested. Reactivity with non-phosphorylated human HMGNs is minimal by ELISA. A BLAST analysis was used to suggest cross reactivity with HMGN proteins from Xenopus species, chicken, mouse, bovine, dog, orangutan, rat and swine based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known. |
| Target Species |
Human, Sheep, Chicken, Canine, Orangutan, Xenopus species |
| Content And Storage |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Isotype |
IgG |
|
Show More
Show Less
|
|