Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGN1/HMG14 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP302946100UL
Description
HMGN1/HMG14 Polyclonal specifically detects HMGN1/HMG14 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HMGN1/HMG14 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
FLJ31471, High mobility group nucleosome-binding domain-containing protein 1, high-mobility group (nonhistone chromosomal) protein 14, high-mobility group nucleosome binding 1, high-mobility group nucleosome binding domain 1, HMG14FLJ27265, MGC104230, MGC117425, non-histone chromosomal protein HMG-14, nonhistone chromosomal protein HMG-14 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HMGN1/HMG14 (NP_004956.5). | |
100 μL | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Phospho Specific, Stem Cell Markers | |
3150 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction