Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGN1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992889
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2889 100 μL
50-199-2890 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2889 Supplier Active Motif Supplier No. 39833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The high-mobility group (HMG) proteins 14 and 17 are abundant chromosomal proteins that bind to nucleosomes and enhance transcription. HMG-14 and HMG-17 also function as architectural elements, which alter the structure of the chromatin fiber and enhance transcription from chromatin templates. HMG-14/17 proteins modify the nucleosomal organization of the 30 nm chromatin fiber and mediate the unfolding of the higher order chromatin structure thereby facilitating access to the underlying DNA sequence. Clustering of architectural elements, such as HMG proteins and linker histone subtypes into distinct domains, may lead to structural and functional heterogeneity along the chromatin fiber. In addition, HMG-14 and HMG-17 have been identified as constitutive components of mouse oocyte and embryonic chromatin that establish a link between the structure of embryonic chromatin and the normal progression of embryonic development.

Specifications

Antigen HMGN1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene HMGN1
Gene Accession No. P05114
Gene Alias BOS_1365; FLJ27265; FLJ31471; high mobility group nucleosomal binding domain 1; high mobility group nucleosome binding domain 1; High mobility group nucleosome-binding domain-containing protein 1; high mobility group protein 14; high-mobility group (nonhistone chromosomal) protein 14; high-mobility group nucleosome binding 1; high-mobility group nucleosome binding domain 1; high-mobility group nucleosome-binding domain-containing protein 1; HMG 14; Hmg14; Hmg-14; HMGN 1; HMGN1; MGC104230; MGC117425; Non-histone chromosomal protein HMG-14; nonhistone chromosomal protein HMG-14
Gene Symbols HMGN1
Host Species Rabbit
Immunogen This HMGN1 antibody was raised against a peptide within the C-terminal region of human HMGN1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.