Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585030 Supplier Invitrogen™ Supplier No. PA585030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKRKAKGDAK GDKAKVKDEP Q Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 90%.

The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGN2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGN2
Gene Accession No. P05204
Gene Alias high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; high mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17
Gene Symbols HMGN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HMGN2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.