Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hmgn2 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993339
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3339 10 μL
50-199-3338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3339 Supplier Active Motif Supplier No. 61354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.

Specifications

Antigen Hmgn2
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene HMGN2
Gene Accession No. P05204
Gene Alias high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; High mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17
Gene Symbols HMGN2
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human Hmgn2.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3151
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.