Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGN4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727372 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7372 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7372 Supplier Proteintech Group Inc Supplier No. 116861AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGN4, also named as High mobility group nucleosome-binding domain-containing protein 4, is a 90 amino acid protein, which belongs to the HMGN family. HMGN4 as a DNA-binding protein localizes in the nucleus. The calculated molecular weight of HMGN4 is 10 kDa, but the modified HMGN4 protein is about 15 kDa.

Specifications

Antigen HMGN4
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HMGN4
Gene Accession No. O00479
Gene Alias HMG17L3, HMGN4, NHC
Gene Symbols HMGN4
Host Species Rabbit
Immunogen HMGN4 Fusion Protein Ag2298
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10473
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.