Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551397 Supplier Invitrogen™ Supplier No. PA551397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSAMLVPVTP EVKPKRTSSS RKMKTKSDMM EENIDTSAQA VAETKQEAVV EEDYNENAKN GEAKITEAPA SEKEIVEVKE ENIEDATEKG GEKKEAVAAE VK Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 57%.

This gene encodes a nuclear protein with similarities to the high mobility group proteins, HMG14 and HMG17, which suggests that this protein may function as a nucleosomal binding and transcriptional activating protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGN5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGN5
Gene Accession No. P82970
Gene Alias high mobility group nucleosome binding domain 5; high mobility group nucleosome-binding domain-containing protein 5; high-mobility group nucleosome binding domain 5; HMGN5; NBP-45; NSBP1; nucleosomal binding protein 1; nucleosome-binding protein 1
Gene Symbols HMGN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HMGN5. Recombinant protein control fragment (Product #RP-91963).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79366
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.