Learn More
HMGN5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304206AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
NSBP1 binds specifically to nuclear core particles and acts as a transcriptional activator.Specifications
HMGN5 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P82970 | |
HMGN5 | |
Between 232 and 282 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HMGN5 | |
high-mobility group nucleosome binding domain 5, NBP-45, NSBP1, nucleosomal binding protein 1, nucleosome-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79366 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.