Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGXB4 Monoclonal Antibody (OTI1G5), TrueMAB™, OriGene
SDP

Catalog No. 501678988
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8988 Supplier OriGene Supplier No. TA801841
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

High mobility group (HMG) proteins are nonhistone chromosomal proteins.

Specifications

Antigen HMGXB4
Applications Western Blot
Classification Monoclonal
Clone OTI1G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HMGXB4
Gene Accession No. Q9UGU5
Gene Alias HMG2L1, HMGBCG, THC211630
Gene Symbols HMGXB4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-348 of human HMGXB4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10042
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.