Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGXB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551448 Supplier Invitrogen™ Supplier No. PA551448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDDSVKKEDC FDGDHTFEDI GLAAGRSQRE KKRSYKDFLR EEEEIAAQVR NSSKKKLKDS ELYFLGTDTH KKKRKHSSDD YYYGDISSLE SSQKKKKKSS PQSTDTAMDL LKAITSPLAA GSKPSKKTGE KSSGSSSHSE S Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

High mobility group (HMG) proteins are nonhistone chromosomal proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGXB4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGXB4
Gene Accession No. Q9UGU5
Gene Alias high mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; HMG2L1; HMGBCG; HMG-box containing 4; HMGXB4; Protein HMGBCG; Protein THC211630; THC211630
Gene Symbols HMGXB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HMGXB4. Recombinant protein control fragment (Product #RP-91373).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10042
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.