Learn More
HMMR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HMMR |
---|---|
Concentration | 0.33 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0506
![]() ![]() |
Proteintech Group Inc
158201AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Antibodies raised against RHAMM have been reported to recognize proteins of 56, 66 and 70 kDa in the supernatant of murine 3T3 fibroblasts. Some of the isoforms detectable by anti-RHAMM antibodies, including the 52 kDa form and a 48-49 kDa form found in 1
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
HMMR | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O75330, P97779, Q00547 | |
15366, 25460, 3161 | |
Peptide | |
Primary | |
-20°C | |
HMMR |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD168, HMMR, IHABP, RHAMM | |
HMMR | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.