Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMOX1 Monoclonal Antibody (GT1334)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531557
Description
Keep as concentrated solution. Predicted reactivity: Mouse (81%), Rat (80%), Dog (85%), Pig (82%), Bovine (83%). Positive Control: HeLa(500 μM CoCl2 treatment for 24 hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
Specifications
HMOX1 | |
Monoclonal | |
2.09 mg/mL | |
PBS with no preservative | |
P09601 | |
HMOX1 | |
Recombinant protein encompassing a sequence within the center region of human Heme Oxygenase 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
GT1334 | |
Unconjugated | |
HMOX1 | |
bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein | |
Mouse | |
Protein A | |
RUO | |
3162 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction