Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMOX1 Monoclonal Antibody (GTS-1), Invitrogen™

Catalog No. PIMA125475
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125475 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125475 Supplier Thermo Scientific Supplier No. MA125475

Mouse Monoclonal Antibody

MA1-25475 will not react with rabbit.

Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMOX1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GTS-1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 1% BSA and no preservative; pH 7.4
Gene HMOX1
Gene Accession No. P06762, P09601, P14901
Gene Alias bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein
Gene Symbols HMOX1
Host Species Mouse
Immunogen Rat heme oxygenase 1.
Purification Method purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15368, 24451, 3162
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.