Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMX2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1793720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1793720 20 μL
NBP179371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1793720 Supplier Novus Biologicals Supplier No. NBP17937120UL

Rabbit Polyclonal Antibody

HMX2 Polyclonal specifically detects HMX2 in Human samples. It is validated for Western Blot.

Specifications

Antigen HMX2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_005510
Gene Alias H6 family homeobox 2, H6 homeo box 2, H6L, homeo box (H6 family) 2, homeobox (H6 family) 2, Homeobox protein H6 family member 2, homeobox protein HMX2, NKX5-2
Gene Symbols HMX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HMX2The immunogen for this antibody is HMX2. Peptide sequence SPSHSDFKEEKERLLPAGSPSPGSERPRDGGAERQAGAAKKKTRTVFSRS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3167
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.