Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560346
Description
Immunogen sequence: ELEAANMAHA SAQTLVSMPL VFRDSSLLRV PVPRSLAFPA PLYYPGSNLS ALPLYNLYNK LDY Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Homeobox genes represent a class of transcription factors that play key roles in the regulation of embryogenesis and development. Here we report the identification of a homeobox-containing gene family that is highly conserved at both the nucleotide and amino acid levels in a diverse number of species. These species encompass both vertebrate and invertebrate phylogenies, ranging from Homo sapiens to Drosophila melanogaster.
Specifications
HMX2 | |
Polyclonal | |
Unconjugated | |
HMX2 | |
H6 family homeobox 2; H6 homeo box 2; H6 homeobox 2; H6L; HMX2; homeo box (H6 family) 2; homeobox protein H6 family member 2; Homeobox protein HMX2; homeobox protein Nkx-5.2; MGC32455; NK-related homeobox; Nkx-5.2; Nkx5-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2RU54 | |
HMX2 | |
Recombinant protein corresponding to Human HMX2. Recombinant protein control fragment (Product #RP-99973). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction