Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMX2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981724
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1724 Supplier Bioss Supplier No. BS11853R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Homeobox genes represent a class of transcription factors that play key roles in the regulation of embryogenesis and development. Here we report the identification of a homeobox-containing gene family that is highly conserved at both the nucleotide and amino acid levels in a diverse number of species. These species encompass both vertebrate and invertebrate phylogenies, ranging from Homo sapiens to Drosophila melanogaster.

Specifications

Antigen HMX2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HMX2
Gene Accession No. A2RU54
Gene Alias H6 family homeobox 2; H6 homeo box 2; H6 homeobox 2; H6L; Hmx2; homeo box (H6 family) 2; Homeobox protein H6 family member 2; Homeobox protein HMX2; Homeobox protein Nkx-5.2; MGC32455; NK-related homeobox; Nkx-5.2; Nkx5-2
Gene Symbols HMX2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HMX2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3167
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.