Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569500
Description
This target displays homology in the following species: Cow: 91%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%; Yeast: 90%; Zebrafish: 75%.
Hmx3 is a transcription factor involved in specification of neuronal cell types and which is required for inner ear and hypothalamus development. This protein binds to the 5'-CAAGTG-3' core sequence. It controls semicircular canal formation in the inner ear and is also required for hypothalamic/pituitary axis of the CNS.
Specifications
HMX3 | |
Polyclonal | |
Unconjugated | |
HMX3 | |
H6 family homeobox 3; H6 family homeobox 3a; H6 homeo box 3; H6 homeobox 3; hmx3; hmx3a; homeo box (H6 family) 3; homeobox (H6 family) 3; homeobox protein H6 family member 3; Homeobox protein HMX3; homeobox protein HMX3; LOW QUALITY PROTEIN: homeobox protein HMX3; homeobox protein Nkx-5.1; Nkx5.1; Nkx-5.1; Nkx5-1; RGD1559927; transcription factor; zgc:109845 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P42581 | |
HMX3 | |
synthetic peptide corresponding to a region of mouse HMX3 (aa 151-200). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction