Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572623
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572623 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572623 Supplier Invitrogen™ Supplier No. PA572623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMX3 Polyclonal Antibody for Western Blot

Hmx3 is a transcription factor involved in specification of neuronal cell types and which is required for inner ear and hypothalamus development. This protein binds to the 5'-CAAGTG-3' core sequence. It controls semicircular canal formation in the inner ear and is also required for hypothalamic/pituitary axis of the CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMX3
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hmx3a
Gene Accession No. Q504H8
Gene Alias H6 family homeobox 3; H6 family homeobox 3a; H6 homeo box 3; H6 homeobox 3; hmx3; hmx3a; homeo box (H6 family) 3; homeobox (H6 family) 3; homeobox protein H6 family member 3; Homeobox protein HMX3; homeobox protein HMX3; LOW QUALITY PROTEIN: homeobox protein HMX3; homeobox protein Nkx-5.1; Nkx5.1; Nkx-5.1; Nkx5-1; RGD1559927; transcription factor; zgc:109845
Gene Symbols hmx3a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 132-160 amino acids from the Central region of DANRE hmx3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60310
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.