Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144698 Supplier Invitrogen™ Supplier No. PA5144698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTTTFKGVDP NSRNSSRVLR PPGGGSNFSL GFDEPTEQPV RKNKMASNIF GTPEENQASW AKSAGAKSSG GREDLESSGL QRRNSSEASS GDFLDLKGEG DIHENVDTDL PGSLGQSEEK PVPAAPVPSP VAPAPVPSRR NP Antigen sequence identity: Mouse 89%; Rat 87%.

HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JPT1
Gene Accession No. Q9UK76
Gene Alias Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed
Gene Symbols JPT1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 3-144. Recombinant protein control fragment (Product #RP-109753).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51155
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.