Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HN1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559774
Description
Immunogen sequence: FGSPVTATSR LAHPNKPKDH VFLCEGEEPK SDLKAARSIP AGAEPGEKGS ARKAGPAKEQ EPMPTVDSHE PRLGPRPRSH NKVLNPPGGK SSISF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 79%.
Hematological and neurological expressed 1-like protein (HN1L) is expressed in liver, kidney, prostate, testis and uterus.
Specifications
HN1L | |
Polyclonal | |
Unconjugated | |
JPT2 | |
2810430B18Rik; AL022629; C16orf34; CRAMP_1 like; D17Ertd441e; hematological and neurological expressed 1-like; hematological and neurological expressed 1-like protein; hematopoietic- and neurologic-expressed sequence 1-like; HN1 like; HN1L; HN1-like protein; Jpt2; Jupiter microtubule associated homolog 2; L11; RGD1305117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90861 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H910 | |
JPT2 | |
Recombinant protein corresponding to Human HN1L. Recombinant protein control fragment (Product #RP-98812). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction