Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HN1L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8796 Supplier Bioss Supplier No. BS9070R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hematological and neurological expressed 1-like protein (HN1L) is expressed in liver, kidney, prostate, testis and uterus.

Specifications

Antigen HN1L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene JPT2
Gene Accession No. Q9H910
Gene Alias 2810430B18Rik; AL022629; C16orf34; CRAMP_1 like; D17Ertd441e; hematological and neurological expressed 1-like; Hematological and neurological expressed 1-like protein; hematopoietic- and neurologic-expressed sequence 1-like; HN1 like; HN1L; HN1-like protein; JPT2; Jupiter microtubule associated homolog 2; L11; RGD1305117
Gene Symbols JPT2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HN1L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90861
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.