Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (3B11NB) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | HNF-3 alpha/FoxA1 |
---|---|
Clone | 3B11NB |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2-4 ug/ml, Flow Cytometry 25 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:25 - 1:50, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP245269SS
![]() |
Novus Biologicals
NBP245269SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP245269
![]() |
Novus Biologicals
NBP245269 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
HNF-3 alpha/FoxA1 Monoclonal specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HNF-3 alpha/FoxA1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
FOXA1 | |
IgG2b κ | |
Protein G purified |
3B11NB | |
Western Blot 2-4 ug/ml, Flow Cytometry 25 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:25 - 1:50, Immunohistochemistry-Paraffin 5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P55317 | |
3169 | |
Partial recombinant human FOXA1 protein (between amino acids 350-500) [Uniprot: P55317]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title