Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (FOXA1/1241), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | HNF-3 alpha/FoxA1 |
---|---|
Clone | FOXA1/1241 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25321120
![]() |
Novus Biologicals
NBP25321120UG |
20 μg |
Each for $268.00
|
|
|||||
NB253211100
![]() |
Novus Biologicals
NBP253211100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253211200
![]() |
Novus Biologicals
NBP253211200UG |
200 μg | N/A | N/A | N/A | ||||
Description
HNF-3 alpha/FoxA1 Monoclonal specifically detects HNF-3 alpha/FoxA1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HNF-3 alpha/FoxA1 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Stem Cell Markers | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
3169 | |
Recombinant human FOXA1 protein fragment (aa372-472) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
FOXA1/1241 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
FOXA1 | |
IgG2b κ | |
Protein A or G purified | |
49 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title