Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-3 alpha / FOXA1 Monoclonal Antibody (2F83), Active Motif™
SDP

Mouse Monoclonal Antibody

$156.65 - $638.65

Specifications

Antigen HNF-3 alpha / FOXA1
Clone 2F83
Concentration 1 mg/mL
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2894
SDP
Encompass
View Documents
Active Motif
39838
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-2893
SDP
Encompass
View Documents
Active Motif
39837
100 μg
Each for $638.65
Only null left
Add to Cart
 
Description

Description

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Specifications

Specifications

HNF-3 alpha / FOXA1
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; Forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; Hepatocyte nuclear factor 3-alpha; HNF3A; Hnf-3a; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2
FOXA1
IgG
Protein G
Antibody
2F83
ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Unconjugated
Mouse
Human, Mouse
P35582, P55317
15375, 3169
This HNF-3 alpha / FOXA1 antibody was raised against a recombinant protein corresponding to amino acids 7-103 of rat HNF-3 alpha /FOXA1.
Primary
-20°C, Avoid Freeze/Thaw Cycles
FOXA1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.