Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-3 beta/FoxA2 Antibody (2141A), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2400MFV450
Description
HNF-3 beta/FoxA2 Monoclonal antibody specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| HNF-3 beta/FoxA2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2141A | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B | |
| Human/Mouse/Rat HNF-3 beta/FoxA2 synthetic peptide, Accession # Q9Y261 | |
| RUO | |
| 3170 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction