Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-3 beta/FoxA2 Antibody (OTI3C10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202088
Description
HNF-3 beta/FoxA2 Monoclonal specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HNF-3 beta/FoxA2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:50 | |
FOXA2 | |
Recombinant protein expressed in E.coli corresponding to amino acids 300-458 of human FOXA2 | |
Affinity Purified | |
RUO | |
3170 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI3C10 | |
Unconjugated | |
forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B | |
Mouse | |
48.1 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction