Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-4 alpha/NR2A1 Antibody (OTI2H2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270905AF405
Description
HNF-4 alpha/NR2A1 Monoclonal specifically detects HNF-4 alpha/NR2A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HNF-4 alpha/NR2A1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HNF4A | |
Recombinant protein fragment corresponding to amino acids 1-315 of human HNF-4 alpha/NR2A1 (NP_849180) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2H2 | |
Western Blot, Immunohistochemistry | |
FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14 | |
Mouse | |
Affinity Purified | |
Phospho Specific, Stem Cell Markers | |
3172.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction