Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-4 gamma/NR2A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | HNF-4 gamma/NR2A2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HNF-4 gamma/NR2A2 Polyclonal specifically detects HNF-4 gamma/NR2A2 in Human samples. It is validated for Western Blot.Specifications
HNF-4 gamma/NR2A2 | |
Polyclonal | |
Rabbit | |
GPCR | |
hepatocyte nuclear factor 4, gamma, hepatocyte nuclear factor 4-gamma, HNF-4-gamma, NR2A2Nuclear receptor subfamily 2 group A member 2, NR2A3 | |
HNF4G | |
IgG | |
46 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q14541 | |
3174 | |
Synthetic peptides corresponding to HNF4G(hepatocyte nuclear factor 4, gamma) The peptide sequence was selected from the C terminal of HNF4G (NP_004124). Peptide sequence QDPLTGQTILLGPMSTLVHADQISTPETPLPSPPQGSGQEQYKIAANQAS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title