Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-6/ONECUT1 Antibody (OTI4F12), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270906AF594
Description
HNF-6/ONECUT1 Monoclonal specifically detects HNF-6/ONECUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
HNF-6/ONECUT1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ONECUT1 | |
Recombinant protein fragment corresponding to amino acids 85-198 of human HNF-6/ONECUT1 (NP_004489) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4F12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
Mouse | |
0.1 mL | |
3175 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction