Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF1 alpha Polyclonal Antibody, Huabio

Rabbit Polyclonal Antibody

Supplier:  HUABIO RESEARCH INC HA500285

Encompass
Encompass_Preferred

Catalog No. 50-254-4557


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

HNF1 alpha Polyclonal antibody specifically detects HNF1 alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

HNF1 alpha
Polyclonal
Unconjugated
Hnf1a
AI323641; albumin proximal factor; hepatic nuclear factor 1; hepatic nuclear factor-1-alpha; hepatocyte nuclear factor 1; hepatocyte nuclear factor 1-alpha; Hepatocyte nuclear factor 1-alpha-A; HNF1; Hnf-1; hnf-1 alpha; HNF1 homeobox A; HNF1 homeobox A L homeolog; HNF1 homeobox A S homeolog; HNF1[a]; hnf1a; Hnf-1a; hnf1-a; hnf1a.L; hnf1a.S; HNF-1A-A; hnf1a-a; hnf1a-b; Hnf1alpha; HNF-1-alpha; HNF1-alpha; HNF-1-alpha-A; I79_010330; IDDM20; interferon production regulator factor; LFB1; LF-B1; LF-B1 hepatic nuclear factor (HNF1): albumin proximal factor also TCF1; LF-B1, hepatic nuclear factor (HNF1): albumin proximal factor, also TCF1; lfb1a; LFB1a1; Liver-specific transcription factor LF-B1; MODY3; TCF1; Tcf-1; TCF-1-A; tcf1-a; transcription factor 1; Transcription factor 1 hepatic; Transcription factor 1, hepatic; transcription factor 1, transcription factor LFB1 /HNF1; transcription factor 1-A; XELAEV_18010473mg; XLFB1; XLFB1a; XLFB1b
Recombinant protein within human HNF1 alpha aa 100-300/631.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
P15257, P20823, P22361
Rabbit
Antigen affinity chromatography
RUO
21405, 24817, 6927
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.