Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF1 Antibody (HNF1A/2087), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279911AF750
Description
HNF1 Monoclonal specifically detects HNF1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HNF1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HNF1A | |
| Recombinant fragment (around aa 214-339) of human HNF1 protein (exact sequence is proprietary) (Uniprot: P20823) | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| Human | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HNF1A/2087 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic; LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction