Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116983 Supplier Invitrogen™ Supplier No. PA5116983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, LO2 Immunogen sequence: LGEPGPYLLA GEGPLDKGES CGGGRGELAE LPNGLGETRG SEDETDDDGE DFTPPILKEL ENLSPEEAAH QKAVVETLLQ EDPWRVAKMV KSYLQQHNIP Q.

This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Hnf1a
Gene Accession No. P15257, P20823, P22361
Gene Alias AI323641; albumin proximal factor; hepatic nuclear factor 1; hepatic nuclear factor-1-alpha; hepatocyte nuclear factor 1; hepatocyte nuclear factor 1-alpha; Hepatocyte nuclear factor 1-alpha-A; HNF1; Hnf-1; hnf-1 alpha; HNF1 homeobox A; HNF1 homeobox A L homeolog; HNF1 homeobox A S homeolog; HNF1[a]; hnf1a; Hnf-1a; hnf1-a; hnf1a.L; hnf1a.S; HNF-1A-A; hnf1a-a; hnf1a-b; Hnf1alpha; HNF-1-alpha; HNF1-alpha; HNF-1-alpha-A; I79_010330; IDDM20; interferon production regulator factor; LFB1; LF-B1; LF-B1 hepatic nuclear factor (HNF1): albumin proximal factor also TCF1; LF-B1, hepatic nuclear factor (HNF1): albumin proximal factor, also TCF1; lfb1a; LFB1a1; Liver-specific transcription factor LF-B1; MODY3; TCF1; Tcf-1; TCF-1-A; tcf1-a; transcription factor 1; Transcription factor 1 hepatic; Transcription factor 1, hepatic; transcription factor 1, transcription factor LFB1 /HNF1; transcription factor 1-A; XELAEV_18010473mg; XLFB1; XLFB1a; XLFB1b
Gene Symbols Hnf1a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-130 of human HNF1a (NP_0005366).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21405, 24817, 6927
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.