Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF1BB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572652
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572652 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572652 Supplier Invitrogen™ Supplier No. PA572652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HNF1BB Polyclonal Antibody for Western Blot

Transcription factor, probably binds to the inverted palindrome 5'-GTTAATNATTAAC-3'. Acts downstream of hnf1ba but is not required for induction of rhombomere r5/r6 gene expression in the hindbrain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF1BB
Applications Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hnf1bb
Gene Accession No. Q8UW00
Gene Alias hepatocyte nuclear factor 1-beta-B; Hepatocyte nuclear factor 1-beta-like; Hepatocyte nuclear factor 1-gamma; HNF1 homeobox Bb; hnf1bb; HNF-1-beta-B; hnf1bl; hnf1g; hnf1gamma; HNF1-gamma; liver-enriched atypical homeobox-containing transcription factor; liver-enriched atypical homeobox-containing transcription factor; transcription factor 2 like protein; wu:fc23f06
Gene Symbols hnf1bb
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 212-245 amino acids from the Central region of DANRE hnf1bb.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 324283
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.