Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF4A/HNF4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100879 Supplier Invitrogen™ Supplier No. PA5100879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HNF4 alpha /gamma.

The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal dominant non-insulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants encoding several different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF4A/HNF4G
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hnf4g
Gene Accession No. P22449, P41235, P49698, Q14541, Q9WUU6
Gene Alias alpha transcription factor 4; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4 gamma; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4, gamma; hepatocyte nuclear factor 4-alpha; Hepatocyte nuclear factor 4-gamma; Hnf4; HNF-4; HNF4 alpha; HNF4A; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; Hnf4g; HNF-4-gamma; MODY; MODY1; Nr2a1; Nr2a2; NR2A21; NR2A3; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; Nuclear receptor subfamily 2 group A member 2; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4
Gene Symbols Hnf4a, Hnf4g
Host Species Rabbit
Immunogen A synthesized peptide derived from human HNF4A(Accession P41235), corresponding to amino acid residues A287-L337.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15378, 25735, 30942, 3172, 3174, 365744
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.