Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF4A Monoclonal Antibody (H6939)
GREENER_CHOICE

Catalog No. 417800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-780-0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-780-0 Supplier Invitrogen™ Supplier No. 417800
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognizes human HNF4 alpha 7- 9 and cross reacts with mouse and rat HNF4 alpha 7- 9.

Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF4A
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H6939
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Physiological saline with 0.1% sodium azide
Gene HNF4A
Gene Accession No. P22449, P41235, P49698
Gene Alias alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4
Gene Symbols HNF4A
Host Species Mouse
Immunogen Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant human HNF4--alpha (1-16 aa).
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15378, 25735, 3172
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.