Learn More
HNF4A Mouse anti-Human, Clone: OTI8B8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806576

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.Specifications
HNF4A | |
Monoclonal | |
Unconjugated | |
HNF4A | |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8B8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P41235 | |
HNF4A | |
Human recombit protein fragment corresponding to amino acids 1-315 of human HNF4A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.