Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF4A Mouse anti-Human, Unconjugated, Clone: 4E2, Abnova™

Catalog No. 89339160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-160 Supplier Abnova Corporation Supplier No. H00003172M04J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

HNF4A Monoclonal antibody specifically detects HNF4A in Human samples. It is validated for WB-Ce, IF, S-ELISA, ELISA, WB-Re, WB-Tr, RNAi-Ab.

Specifications

Antigen HNF4A
Applications Western Blot, Immunofluorescence, ELISA, KnockDown
Classification Monoclonal
Clone 4E2
Conjugate Unconjugated
Description hepatocyte nuclear factor 4, alpha
Gene HNF4A
Gene Accession No. NM_000457.4
Gene Alias FLJ39654, HNF4, HNF4a7, HNF4a8, HNF4a9, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
Host Species Mouse
Immunogen HNF4A (NP_000448.3, 324 a.a. ∼ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3172
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.