Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HNF4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120781
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, HT-29. The target is usually found in the following locations: Nucleus. Immunogen sequence: EHLLLGATKR SMVFKDVLLL GNDYIVPRHC PELAEMSRVS IRILDELVLP FQELQIDDNE YAYLKAIIFF DPDAKGLSDP GKIKRLRSQV QVSLEDYIND.
Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.
Specifications
| HNF4A | |
| Polyclonal | |
| Unconjugated | |
| HNF4A | |
| alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3172 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P41235 | |
| HNF4A | |
| A synthetic peptide corresponding to a sequence within amino acids 226-325 of human HNF4A (NP_8491801). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction