Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582189 Supplier Invitrogen™ Supplier No. PA582189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WQMIEQIQFV KLFGMVKIDN LLQEMLLGGA SNDGSHLHHP MHPHLSQDPL TGQTILLGPM STLVHADQIS TPETPLPSPP QGSGQEQYKI AANQASVISH QHLSKQKQ Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

HNF4G is a transcription factor. It has a lower transcription activation potential than HNF4-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF4G
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HNF4G
Gene Accession No. Q14541
Gene Alias hepatocyte nuclear factor 4 gamma; hepatocyte nuclear factor 4, gamma; hepatocyte nuclear factor 4-gamma; HNF4 gamma; HNF-4 gamma; HNF4-beta; Hnf4g; Hnf4gamma; HNF-4-gamma; NR2A2; NR2A3; Nuclear receptor subfamily 2 group A member 2
Gene Symbols HNF4G
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HNF4G. Recombinant protein control fragment (Product #RP-89212).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3174
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.