Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590904 Supplier Invitrogen™ Supplier No. PA590904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASNDGSHLHH PMHPHLSQDP LTGQTILLGP MSTLVHADQI STPETPLPSP PQGSGQEQYK IAANQASVIS HQHLSKQKQL; Positive Samples: HT-29, Mouse pancreas, Rat pancreas, Rat kidney; Cellular Location: Nucleus.

HNF4G is a transcription factor. It has a lower transcription activation potential than HNF4-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF4G
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene HNF4G
Gene Accession No. Q14541, Q9WUU6
Gene Alias hepatocyte nuclear factor 4 gamma; hepatocyte nuclear factor 4, gamma; hepatocyte nuclear factor 4-gamma; HNF4 gamma; HNF-4 gamma; HNF4-beta; Hnf4g; Hnf4gamma; HNF-4-gamma; NR2A2; NR2A3; Nuclear receptor subfamily 2 group A member 2
Gene Symbols HNF4G
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 366-445 of human HNF4G (NP_0041244?).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30942, 3174, 365744
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.