Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HNF4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117990 Supplier Invitrogen™ Supplier No. PA5117990
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HNF4G Polyclonal Antibody for Western Blot, IP

HNF4G is a transcription factor. It has a lower transcription activation potential than HNF4-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF4G
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene HNF4G
Gene Accession No. Q9WUU6
Gene Alias hepatocyte nuclear factor 4 gamma; hepatocyte nuclear factor 4, gamma; hepatocyte nuclear factor 4-gamma; HNF4 gamma; HNF-4 gamma; HNF4-beta; Hnf4g; Hnf4gamma; HNF-4-gamma; NR2A2; NR2A3; Nuclear receptor subfamily 2 group A member 2
Gene Symbols HNF4G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Mouse HNF-4 gamma.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30942
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.