Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HNMT Monoclonal Antibody (432CT31.4.2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537557
Description
HNMT Monoclonal Antibody for Western Blot
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.
Specifications
HNMT | |
Monoclonal | |
Unconjugated | |
Hnmt | |
1500031F01Rik; AI788969; histamine N-methyltransferase; Hmt; Hnmt; HNMT-S1; HNMT-S2; MRT51 | |
Mouse | |
Ammonium Sulfate Precipitation | |
RUO | |
3176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
432CT31.4.2 | |
PBS with 0.09% sodium azide; pH 7.4 | |
P50135 | |
Hnmt | |
purified His-tagged HNMT protein (fragment). | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction