Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HNMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557288
Description
Immunogen sequence: TSDDLTQMLD NLGLKYECYD LLSTMDISDC FIDGNENGDL LWDFLTETCN FNATAPPDLR AELGKDLQEP EFSAKKEGKV LFNNTLSFIV IEA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.
Specifications
| HNMT | |
| Polyclonal | |
| Unconjugated | |
| Hnmt | |
| 1500031F01Rik; AI788969; histamine N-methyltransferase; Hmt; Hnmt; HNMT-S1; HNMT-S2; MRT51 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3176 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50135 | |
| Hnmt | |
| Recombinant protein corresponding to Human HNMT. Recombinant protein control fragment (Product #RP-102666). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction