Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP A1 Antibody (4B10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100672H
Description
hnRNP A1 Monoclonal specifically detects hnRNP A1 in Human, Mouse, Bovine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.Specifications
hnRNP A1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HNRNPA1 | |
Full length partially purified human hnRNP A1. [UniProt# P09651] | |
0.1 mL | |
Primary | |
Does not react with hnRNP A2 or B | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry, KnockDown | |
4B10 | |
Western Blot, ELISA, Immunohistochemistry, Knockdown Validated | |
Helix-destabilizing protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein core protein A1, hnRNP A1, hnRNP core protein A1, hnRNPA1, hnRNP-A1, HNRPA1MGC102835, nuclear ribonucleoprotein particle A1 protein, single-strand DNA-binding protein UP1, Single-strand RNA-binding protein | |
Mouse | |
Protein G purified | |
Core ESC Like Genes, Stem Cell Markers | |
3178 | |
Human, Mouse, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction