Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA519431
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519431 Supplier Thermo Scientific Supplier No. PA519431

Rabbit Polyclonal Antibody

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse, rat and macaque monkey based on sequence homology. Store antibody at 4oC for 1-2 weeks. For long-term storage, store at -20oC.

HnRNP A1 belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre mRNAs in the nucleus and appear to influence pre mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. hnRNP A1 has two repeats of quasi RRM domains that bind to RNAs. It is one of the most abundant core proteins of hnRNP complexes and it is localized to the nucleoplasm. This protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re imported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Hnrnpa1
Gene Accession No. P09651
Gene Alias ALS19; ALS20; Fli-2; Hdp; HDP-1; Helix-destabilizing protein; heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP A1-like 3; hnRNP core protein A1; hnRNP core protein A1-like 3; hnRNP I; Hnrnpa1; hnRNP-A1; HNRNPI; HNRNP-I; Hnrpa1; HNRPA1L3; HNRPA1MGC102835; HNRPI; I79_022927; IBMPFD3; MGC102835; MGC128297 protein; nuclear ribonucleoprotein particle A1 protein; pPTB; PTB; PTB-1; PTB2; PTB3; PTB4; PTB-T; putative heterogeneous nuclear ribonucleoprotein A1-like 3; Putative heterogeneous nuclear ribonucleoprotein A1-like protein 3; ROA1; RP11-78J21.1; single-strand DNA-binding protein UP1; single-strand RNA-binding protein; Single-strand-binding protein; Tis; Topoisomerase-inhibitor suppressed; unwinding protein 1; UP 1; UP1
Gene Symbols Hnrnpa1
Host Species Rabbit
Immunogen Synthetic peptide derived from residues 300 to the C-terminus of Human hnRNP A1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3178
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.