Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP A1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541731 Supplier Invitrogen™ Supplier No. PA541731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSKSASPKEP EQLRKLFIGG LSFETTDESL RSHFEQWGTL TDCVVMRDPN Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Involved in the packaging of pre-mRNA into hnRNP particles, transport of poly(A) mRNA from the nucleus to the cytoplasm and may modulate splice site selection. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP A1L2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HNRNPA1L2
Gene Accession No. Q32P51
Gene Alias heterogeneous nuclear ribonucleoprotein A1-like 2; hnRNP A1-like 2; hnRNP core protein A1-like 2; HNRNPA1L; HNRNPA1L2
Gene Symbols HNRNPA1L2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RP11-78J21.1 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144983
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.