Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Antibody (DP3B3) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 5 publications

$243.50 - $560.50

Specifications

Antigen hnRNP A2B1
Clone DP3B3
Concentration 1.0 mg/mL
Dilution Western Blot 1 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31844323), Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 31844323), KnockDown Validated
Applications Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1206102SS
SDP
View Documents
Novus Biologicals
NB1206102SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB1206102
SDP
View Documents
Novus Biologicals
NB1206102
0.1 mL
Each for $560.50
Only null left
Add to Cart
 
Description

Description

hnRNP A2B1 Monoclonal specifically detects hnRNP A2B1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated.
Specifications

Specifications

hnRNP A2B1
1.0 mg/mL
Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
Unconjugated
Mouse
Human, Mouse, Rat, Bovine, Canine, Chicken, Primate
DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2 hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1
HNRNPA2B1
IgG2a
Protein A purified
Mouse, Human, Chicken, Canine, Rat, Monkey, Bovine and Hamster.
DP3B3
Western Blot 1 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31844323), Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 31844323), KnockDown Validated
Monoclonal
Purified
RUO
P22626, O88569, B2GV69
3181
Bacterially expressed human hnRNP A2 fusion protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.