Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP A2B1 Antibody (DP3B3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1206102MFV610
Description
hnRNP A2B1 Monoclonal antibody specifically detects hnRNP A2B1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, ChIP assay, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP), KnockDown, KnockDownSpecifications
hnRNP A2B1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
Purified |
Western Blot, ChIP Assay, Immunocytochemistry, Immunoprecipitation, ChIP Assay | |
DP3B3 | |
Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated | |
DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2/ hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1 | |
Bacterially expressed human hnRNP A2 fusion protein. | |
0.1 mL | |
Cancer, DNA Repair, DNA replication Transcription Translation and Splicing | |
3181 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction